APO455830062 Apollo Scientific Ltd

1H-Imidazole-2-carboxylic acid

  • MDL number MFCD00270816
  • Cas Number : 16042-25-4
  • Molecular Weight : 112.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 198
Iupac Name 1H-imidazole-2-carboxylic acid
Inchi Code 1S/C4H4N2O2/c7-4(8)3-5-1-2-6-3/h1-2H,(H,5,6)(H,7,8)
Inchi Key KYWMCFOWDYFYLV-UHFFFAOYSA-N

Description

Synonym 2-Carboxy-1H-imidazole
Purity 96%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 198
Product Link https://store.apolloscientific.co.uk/product/1h-imidazole-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1h-imidazole-2-carboxylic-acid
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart