APO455830070 Apollo Scientific Ltd

5-Bromo-2,4-dimethylaniline

  • MDL number MFCD09038481
  • Cas Number : 69383-60-4
  • Molecular Weight : 200.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 96-100
Iupac Name 5-bromo-2,4-dimethylaniline
Inchi Code 1S/C8H10BrN/c1-5-3-6(2)8(10)4-7(5)9/h3-4H,10H2,1-2H3
Inchi Key LRPYBWZQRJDXDB-UHFFFAOYSA-N

Description

Synonym 4-Amino-6-bromo-m-xylene, 5-Bromo-2,4-xylidine
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 96-100
Product Link https://store.apolloscientific.co.uk/product/5-bromo-24-dimethylaniline

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-24-dimethylaniline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart