APO455830072 Apollo Scientific Ltd

4,6-Dichloro-1-methyl-1H-pyrazolo[3,4-d]pyrimidine

  • MDL number MFCD09750213
  • Cas Number : 98141-42-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/46-dichloro-1-methyl-1h-pyrazolo34-dpyrimidine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/46-dichloro-1-methyl-1h-pyrazolo34-dpyrimidine
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart