APO455830078 Apollo Scientific Ltd

N-[2-(Piperazin-1-yl)phenyl]methylsulphonamide hydrochloride

  • MDL number MFCD11869756
  • Cas Number : 199105-19-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 64-68
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=44886

Safety Information

signalword Danger
pcodes P260;P280;P261
MSDS http://www.apolloscientific.co.uk/display_item.php?id=44886
pictograms GHS05;GHS07
hstatements H302;H312;H332;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart