APO455830079 Apollo Scientific Ltd

8-Aza-1-oxaspiro[4.5]decane hydrochloride

  • MDL number MFCD11042294
  • Cas Number : 3970-79-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/8-aza-1-oxaspiro45decane-hydrochloride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/8-aza-1-oxaspiro45decane-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart