APO455830081 Apollo Scientific Ltd

2-Methyl-1,3,7-triazaspiro[4.5]dec-1-en-4-one hydrochloride

  • MDL number MFCD11869758
  • Cas Number : 1214028-87-1
  • Molecular Weight : 203.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-methyl-1,3,7-triazaspiro[4.5]dec-1-en-4-one hydrochloride
Inchi Code 1S/C8H13N3O.ClH/c1-6-10-7(12)8(11-6)3-2-4-9-5-8;/h9,11H,1-5H2,(H,10,12);1H
Inchi Key RGBMOXSPCAGPCC-UHFFFAOYSA-N

Description

Synonym 2-Methyl-4-oxo-1,3,7-triazaspiro[4.5]dec-1-ene hydrochloride
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-methyl-137-triazaspiro45dec-1-en-4-one-hydrochloride

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-methyl-137-triazaspiro45dec-1-en-4-one-hydrochloride
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart