APO455830087 Apollo Scientific Ltd

3-Bromo-4-azaindole

  • MDL number MFCD10697525
  • Cas Number : 23688-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Bromo-1H-pyrrolo[3,2-b]pyridine
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-bromo-4-azaindole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-4-azaindole
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart