APO455830089 Apollo Scientific Ltd

3-Bromo-6-azaindole

  • MDL number MFCD06738910
  • Cas Number : 67058-76-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Bromo-1H-pyrrolo[2,3-c]pyridine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-bromo-6-azaindole

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-6-azaindole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart