APO455830100 Apollo Scientific Ltd

5-Acetylpyrimidine

  • MDL number MFCD09038477
  • Cas Number : 10325-70-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 87-88
Synonym 1-(Pyrimidin-5-yl)ethan-1-one, 5-Ethanoylpyrimidine
Purity 98
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 70/2mm
Product Link https://store.apolloscientific.co.uk/product/5-acetylpyrimidine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-acetylpyrimidine
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart