APO455830127 Apollo Scientific Ltd

6-Bromo-3-methyl-1H-indazole

  • MDL number MFCD08272244
  • Cas Number : 7746-27-2
  • Molecular Weight : 211.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-bromo-3-methyl-1H-indazole
Inchi Code 1S/C8H7BrN2/c1-5-7-3-2-6(9)4-8(7)11-10-5/h2-4H,1H3,(H,10,11)
Inchi Key PUVRYFUBGFMXMW-UHFFFAOYSA-N

Description

Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-bromo-3-methyl-1h-indazole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/6-bromo-3-methyl-1h-indazole
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart