APO455830135 Apollo Scientific Ltd

4-Bromo-2-methylisoindolin-1-one

  • MDL number MFCD11878013
  • Cas Number : 435273-55-5
  • Molecular Weight : 226.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-bromo-2-methyl-1-isoindolinone
Inchi Code 1S/C9H8BrNO/c1-11-5-7-6(9(11)12)3-2-4-8(7)10/h2-4H,5H2,1H3
Inchi Key HNVZVGCFIFFWMV-UHFFFAOYSA-N

Description

Synonym 4-Bromo-2,3-dihydro-2-methyl-1H-isoindol-1-one, 4-Bromo-2,3-dihydro-2-methyl-1-oxo-1H-isoindole
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-2-methylisoindolin-1-one

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-2-methylisoindolin-1-one
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart