APO455830140 Apollo Scientific Ltd

4,6-Dichloropyridine-2-carboxylic acid

  • MDL number MFCD09910267
  • Cas Number : 88912-25-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111-114
Synonym 2-Carboxy-4,6-dichloropyridine, 4,6-Dichloropicolinic acid
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/46-dichloropyridine-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/46-dichloropyridine-2-carboxylic-acid
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart