APO455830142 Apollo Scientific Ltd

Methyl 2-hydroxy-6-methylisonicotinate

  • MDL number MFCD13188667
  • Cas Number : 98491-78-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Methyl 2-hydroxy-6-methylpyridine-4-carboxylate, 2-Hydroxy-4-(methoxycarbonyl)-6-methylpyridine
Purity 95
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/methyl-2-hydroxy-6-methylisonicotinate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-2-hydroxy-6-methylisonicotinate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart