APO455830147 Apollo Scientific Ltd

4-[(Methylamino)methyl]tetrahydro-2H-pyran

  • MDL number MFCD06739009
  • Cas Number : 439081-52-4
  • Molecular Weight : 129.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-methyl(tetrahydro-2H-pyran-4-yl)methanamine
Inchi Code 1S/C7H15NO/c1-8-6-7-2-4-9-5-3-7/h7-8H,2-6H2,1H3
Inchi Key WMBCUXKYKVTJRF-UHFFFAOYSA-N

Description

Synonym N-Methyl-1-(tetrahydro-2H-pyran-4-yl)methylamine, N-Methyl-1-(oxan-4-yl)methylamine
Flash Point 110/15mm
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-methylaminomethyltetrahydro-2h-pyran

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-methylaminomethyltetrahydro-2h-pyran
pictograms GHS05;GHS07
hstatements H302;H314
Availability details for
Quantity
  • Availability
  • Add to Cart