APO455830151 Apollo Scientific Ltd

5-[(Morpholin-4-yl)methyl]thiophene-2-carboxaldehyde

  • MDL number MFCD06803315
  • Cas Number : 893744-01-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90-92
Synonym 2-Formyl-5-[(morpholin-4-yl)methyl]thiophene, 5-[(Morpholin-4-yl)methyl]-2-thenaldehyde
Purity 97
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=45026

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=45026
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart