APO455830152 Apollo Scientific Ltd

Dihydrofuran-3(2H)-one

  • MDL number MFCD07778393
  • Cas Number : 22929-52-8
  • Molecular Weight : 86.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name dihydro-3(2H)-furanone
density 1.1124
Inchi Code 1S/C4H6O2/c5-4-1-2-6-3-4/h1-3H2
Inchi Key JLPJFSCQKHRSQR-UHFFFAOYSA-N

Description

Synonym 3-Oxo-1,4-epoxybutane, Oxolan-3-one
Density 1.1124
Flash Point 139-140
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/dihydrofuran-32h-one

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/dihydrofuran-32h-one
pictograms GHS07
hstatements H302;H315;H319
Availability details for
Quantity
  • Availability
  • Add to Cart