APO455830157 Apollo Scientific Ltd

5-(Aminomethyl)-1H-indazole

  • MDL number MFCD09028157
  • Cas Number : 267413-25-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (1H-Indazol-5-yl)methylamine
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=44936

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=44936
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart