APO455830159 Apollo Scientific Ltd

6-(Bromomethyl)-1H-indazole hydrobromide

  • MDL number MFCD14586465
  • Cas Number : 368426-63-5
  • Molecular Weight : 291.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-(bromomethyl)-1H-indazole hydrobromide
Inchi Code 1S/C8H7BrN2.BrH/c9-4-6-1-2-7-5-10-11-8(7)3-6;/h1-3,5H,4H2,(H,10,11);1H
Inchi Key DTGUIYRXOJYJLA-UHFFFAOYSA-N

Description

Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-bromomethyl-1h-indazole-hydrobromide

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/6-bromomethyl-1h-indazole-hydrobromide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart