APO455830164 Apollo Scientific Ltd

5-Hydroxy-2-methyl-2H-indazole

  • MDL number MFCD12028572
  • Cas Number : 1159511-41-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Methyl-2H-indazol-5-ol
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=45044

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=45044
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart