APO455830170 Apollo Scientific Ltd

4-(Aminomethyl)-1H-indazole

  • MDL number MFCD10696781
  • Cas Number : 944898-72-0
  • Molecular Weight : 147.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (1H-Indazol-4-yl)methylamine
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-aminomethyl-1h-indazole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-aminomethyl-1h-indazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart