APO455830171 Apollo Scientific Ltd

7-(Aminomethyl)-1H-indazole

  • MDL number MFCD10696780
  • Cas Number : 944904-20-5
  • Molecular Weight : 147.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (1H-Indazol-7-yl)methylamine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/7-aminomethyl-1h-indazole

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/7-aminomethyl-1h-indazole
pictograms GHS05;GHS07
hstatements H302;H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart