APO455830172 Apollo Scientific Ltd

4-(Aminomethyl)-1-methyl-1H-indazole hydrochloride

  • MDL number MFCD18803462
  • Cas Number : 1334405-46-7
  • Molecular Weight : 197.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (1-Methyl-1H-indazol-4-yl)methylamine hydrochloride
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-aminomethyl-1-methyl-1h-indazole-hydrochloride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-aminomethyl-1-methyl-1h-indazole-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart