APO455830176 Apollo Scientific Ltd

7-(Aminomethyl)-2-methyl-2H-indazole

  • MDL number MFCD11869761
  • Cas Number : 1144044-67-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (2-Methyl-2H-indazol-7-yl)methylamine
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=44965

Safety Information

signalword Warning
pcodes P280;P301+P312;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=44965
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart