APO455830178 Apollo Scientific Ltd

5-Acetyl-1H-indazole

  • MDL number MFCD11869763
  • Cas Number : 1001906-63-3
  • Molecular Weight : 160.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(1H-indazol-5-yl)ethanone
Inchi Code 1S/C9H8N2O/c1-6(12)7-2-3-9-8(4-7)5-10-11-9/h2-5H,1H3,(H,10,11)
Inchi Key WYLOAPVBIBQLJT-UHFFFAOYSA-N

Description

Synonym 1-(1H-Indazol-5-yl)ethan-1-one, 5-Ethanoyl-1H-indazole
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-acetyl-1h-indazole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-acetyl-1h-indazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart