APO455830181 Apollo Scientific Ltd

4-Acetyl-1-methyl-1H-indazole

  • MDL number MFCD11869766
  • Cas Number : 1159511-23-5
  • Molecular Weight : 174.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(1-methyl-1H-indazol-4-yl)ethanone
Inchi Code 1S/C10H10N2O/c1-7(13)8-4-3-5-10-9(8)6-11-12(10)2/h3-6H,1-2H3
Inchi Key JWKGENUMGZSMLE-UHFFFAOYSA-N

Description

Synonym 1-(1-Methyl-1H-indazol-4-yl)ethan-1-one, 4-Ethanoyl-1-methyl-1H-indazole
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-acetyl-1-methyl-1h-indazole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-acetyl-1-methyl-1h-indazole
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart