APO455830182 Apollo Scientific Ltd

5-Acetyl-1-methyl-1H-indazole

  • MDL number MFCD11869767
  • Cas Number : 1159511-24-6
  • Molecular Weight : 174.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(1-Methyl-1H-indazol-5-yl)ethan-1-one, 5-Ethanoyl-1-methyl-1H-indazole
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-acetyl-1-methyl-1h-indazole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-acetyl-1-methyl-1h-indazole
pictograms GHS07
hstatements H302;H312;H315;H319;H332;H335
Availability details for
Quantity
  • Availability
  • Add to Cart