APO455830184 Apollo Scientific Ltd

7-Acetyl-1-methyl-1H-indazole

  • MDL number MFCD11869769
  • Cas Number : 1159511-26-8
  • Molecular Weight : 174.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(1-methyl-1H-indazol-7-yl)ethanone
Inchi Code 1S/C10H10N2O/c1-7(13)9-5-3-4-8-6-11-12(2)10(8)9/h3-6H,1-2H3
Inchi Key XGYYSLYITFEONB-UHFFFAOYSA-N

Description

Synonym 1-(1-Methyl-1H-indazol-7-yl)ethan-1-one, 7-Ethanoyl-1-methyl-1H-indazole
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/7-acetyl-1-methyl-1h-indazole

Safety Information

signalword Warning
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/7-acetyl-1-methyl-1h-indazole
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart