APO455830185 Apollo Scientific Ltd

4-Acetyl-2-methyl-2H-indazole

  • MDL number MFCD11869770
  • Cas Number : 1159511-27-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(2-Methyl-2H-indazol-4-yl)ethan-1-one, 4-Ethanoyl-2-methyl-2H-indazole
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=44974

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=44974
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart