APO455830186 Apollo Scientific Ltd

6-Acetyl-2-methyl-2H-indazole

  • MDL number MFCD11869772
  • Cas Number : 1159511-29-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(2-Methyl-2H-indazol-6-yl)ethan-1-one, 6-Ethanoyl-2-methyl-2H-indazole
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=44976

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=44976
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart