APO455830188 Apollo Scientific Ltd

5-Bromo-2-nitrobenzonitrile

  • MDL number MFCD10699674
  • Cas Number : 89642-50-2
  • Molecular Weight : 227.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 116-118
Iupac Name 5-bromo-2-nitrobenzonitrile
Inchi Code 1S/C7H3BrN2O2/c8-6-1-2-7(10(11)12)5(3-6)4-9/h1-3H
Inchi Key LCNNOEPOXHHUQG-UHFFFAOYSA-N

Description

Synonym 4-Bromo-2-cyanonitrobenzene
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 116-118
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-nitrobenzonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-nitrobenzonitrile
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart