APO455830189 Apollo Scientific Ltd

5-Bromo-2-nitrophenol

  • MDL number MFCD03095027
  • Cas Number : 27684-84-0
  • Molecular Weight : 218.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 40-41
Iupac Name 5-bromo-2-nitrophenol
Inchi Code 1S/C6H4BrNO3/c7-4-1-2-5(8(10)11)6(9)3-4/h1-3,9H
Inchi Key DTWHNSNSUBKGTC-UHFFFAOYSA-N

Description

Synonym 4-Bromo-2-hydroxynitrobenzene
Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 40-41
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-nitrophenol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-nitrophenol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart