APO455830190 Apollo Scientific Ltd

2-Amino-5-bromophenol

  • MDL number MFCD03095028
  • Cas Number : 38191-34-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-140(dec.)
Synonym 4-Bromo-2-hydroxyaniline
Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-amino-5-bromophenol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-5-bromophenol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart