APO455830194 Apollo Scientific Ltd

2-Chloro-4,6-dimethylpyridine

  • MDL number MFCD08277279
  • Cas Number : 30838-93-8
  • Molecular Weight : 141.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-chloro-4,6-dimethylpyridine
Inchi Code 1S/C7H8ClN/c1-5-3-6(2)9-7(8)4-5/h3-4H,1-2H3
Inchi Key COVGXNSRDOYAJD-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloro-46-dimethylpyridine

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-46-dimethylpyridine
pictograms GHS07
hstatements H302+H312+H332;H319;H315;H335
Availability details for
Quantity
  • Availability
  • Add to Cart