APO455830196 Apollo Scientific Ltd

3-Amino-2-bromo-4-methylpyridine

  • MDL number MFCD04112497
  • Cas Number : 126325-50-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 78-80
Synonym 2-Bromo-4-methylpyridin-3-amine
Purity 97
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-amino-2-bromo-4-methylpyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-amino-2-bromo-4-methylpyridine
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart