APO455830201 Apollo Scientific Ltd

5-Amino-2-methoxypyrimidine

  • MDL number MFCD08234878
  • Cas Number : 56621-89-7
  • Molecular Weight : 125.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 119-120
Iupac Name 2-methoxy-5-pyrimidinamine
Inchi Code 1S/C5H7N3O/c1-9-5-7-2-4(6)3-8-5/h2-3H,6H2,1H3
Inchi Key HKHRENFWPKWVML-UHFFFAOYSA-N

Description

Synonym 2-Methoxypyrimidin-5-amine, 5-Amino-2-methoxy-1,3-diazine
Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 119-120
Product Link https://store.apolloscientific.co.uk/product/5-amino-2-methoxypyrimidine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-amino-2-methoxypyrimidine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart