APO455830202 Apollo Scientific Ltd

2-Amino-4-ethyl-6-hydroxypyrimidine

  • MDL number MFCD10698994
  • Cas Number : 5734-66-7
  • Molecular Weight : 139.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 249-251
Synonym 2-Amino-6-ethylpyrimidin-4-ol, 4-Ethyl-6-hydroxypyrimidin-2-amine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-amino-4-ethyl-6-hydroxypyrimidine

Safety Information

signalword Warning
pcodes P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-4-ethyl-6-hydroxypyrimidine
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart