APO455830203 Apollo Scientific Ltd

3-Amino-6-methylpyridazine

  • MDL number MFCD01569899
  • Cas Number : 18591-82-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 223-227
Synonym 6-Methylpyridazin-3-amine, 3-Amino-6-methyl-1,2-diazine
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=45516

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=45516
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart