APO455830208 Apollo Scientific Ltd

3-Amino-6-bromoimidazo[1,2-a]pyridine

  • MDL number MFCD09864901
  • Cas Number : 705262-55-1
  • Molecular Weight : 212.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 145-147
Iupac Name 6-bromoimidazo[1,2-a]pyridin-3-amine
Inchi Code 1S/C7H6BrN3/c8-5-1-2-7-10-3-6(9)11(7)4-5/h1-4H,9H2
Inchi Key PGSBYVUIXGOCGV-UHFFFAOYSA-N

Description

Synonym 6-Bromoimidazo[1,2-a]pyridin-3-amine
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 145-147
Product Link https://store.apolloscientific.co.uk/product/3-amino-6-bromoimidazo12-apyridine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-amino-6-bromoimidazo12-apyridine
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart