APO455830211 Apollo Scientific Ltd

3-Amino-1H-indazole

  • MDL number MFCD00182045
  • Cas Number : 874-05-5
  • Molecular Weight : 133.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 156-158
Iupac Name 3H-indazol-3-amine
Inchi Code 1S/C7H7N3/c8-7-5-3-1-2-4-6(5)9-10-7/h1-4H,(H3,8,9,10)
Inchi Key YDTDKKULPWTHRV-UHFFFAOYSA-N

Description

Synonym 1H-Indazol-3-amine
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 156-158
Product Link https://store.apolloscientific.co.uk/product/3-amino-1h-indazole

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-amino-1h-indazole
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart