APO455830215 Apollo Scientific Ltd

1-Methyl-1H-pyrazole-3-carboxaldehyde

  • MDL number MFCD03419800
  • Cas Number : 27258-32-8
  • Molecular Weight : 110.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Formyl-1-methyl-1H-pyrazole
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 88-90/0.1mm
Product Link https://store.apolloscientific.co.uk/product/1-methyl-1h-pyrazole-3-carboxaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-methyl-1h-pyrazole-3-carboxaldehyde
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart