APO455830216 Apollo Scientific Ltd

3-Bromoimidazo[1,2-b]pyridazine

  • MDL number MFCD09757672
  • Cas Number : 18087-73-5
  • Molecular Weight : 198.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 146-149
Iupac Name 3-bromoimidazo[1,2-b]pyridazine
Inchi Code 1S/C6H4BrN3/c7-5-4-8-6-2-1-3-9-10(5)6/h1-4H
Inchi Key KJQVHOFAWISYDO-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 146-149
Product Link https://store.apolloscientific.co.uk/product/3-bromoimidazo12-bpyridazine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromoimidazo12-bpyridazine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart