APO455830217 Apollo Scientific Ltd

3-(5-Chlorothien-2-yl)-1H-pyrazole

  • MDL number MFCD01569447
  • Cas Number : 166196-61-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Chloro-5-(1H-pyrazol-3-yl)thiophene
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=13943

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=13943
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart