APO455830225 Apollo Scientific Ltd

5-(Pyridin-2-yl)thiophene-2-sulphonohydrazide

  • MDL number MFCD00052124
  • Cas Number : 175202-74-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-[5-(Hydrazinosulphonyl)thien-2-yl]pyridine, 2-(Hydrazinosulphonyl)-5-(pyridin-2-yl)thiophene
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=15699

Safety Information

signalword Warning
pcodes P261;P280;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=15699
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart