APO455830243 Apollo Scientific Ltd

5'-Bromo-2'-chloroacetophenone

  • MDL number MFCD02683851
  • Cas Number : 105884-19-3
  • Molecular Weight : 233.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(5-bromo-2-chlorophenyl)ethanone
Inchi Code 1S/C8H6BrClO/c1-5(11)7-4-6(9)2-3-8(7)10/h2-4H,1H3
Inchi Key FVJMYXDLWAEIKP-UHFFFAOYSA-N

Description

Synonym 1-(5-Bromo-2-chlorophenyl)ethan-1-one
Purity 99%
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-chloroacetophenone

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-chloroacetophenone
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart