APO455830247 Apollo Scientific Ltd

1H-1,2,4-Triazole-5-carboxylic acid

  • MDL number MFCD00186418
  • Cas Number : 4928-87-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Carboxy-1H-1,2,4-triazole, 2H-1,2,4-Triazole-3-carboxylic acid, 1H-1,2,4-Triazole-3-carboxylic acid
Purity 98
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=33378

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=33378
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart