APO455830248 Apollo Scientific Ltd

2-[(Trimethylsilyl)ethynyl]benzonitrile

  • MDL number MFCD09260485
  • Cas Number : 97308-62-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym [(2-Cyanophenyl)ethynyl]trimethylsilane
Purity 98
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 100/0.03mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=33379

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=33379
pictograms GHS02;GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart