APO455830258 Apollo Scientific Ltd

5-Acetyl-2-phenyl-1,3-thiazole

  • MDL number MFCD00244691
  • Cas Number : 10045-50-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 151-153
Synonym 1-(2-Phenyl-1,3-thiazol-5-yl)ethan-1-one
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=33411

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=33411
pictograms GHS07
hstatements H302;H319
Availability details for
Quantity
  • Availability
  • Add to Cart