APO455830260 Apollo Scientific Ltd

3-(5-Bromothien-2-yl)-1H-pyrazole

  • MDL number MFCD00111430
  • Cas Number : 166196-67-4
  • Molecular Weight : 229.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 167
Iupac Name 3-(5-bromo-2-thienyl)-1H-pyrazole
Inchi Code 1S/C7H5BrN2S/c8-7-2-1-6(11-7)5-3-4-9-10-5/h1-4H,(H,9,10)
Inchi Key RBOKZFIVGFLORX-UHFFFAOYSA-N

Description

Synonym 2-Bromo-5-(1H-pyrazol-3-yl)thiophene, 5-(5-Bromothien-2-yl)-1H-pyrazole
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 167
Product Link https://store.apolloscientific.co.uk/product/3-5-bromothien-2-yl-1h-pyrazole

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/3-5-bromothien-2-yl-1h-pyrazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart