APO455830261 Apollo Scientific Ltd

3-(Bromoacetyl)benzoic acid

  • MDL number MFCD03789105
  • Cas Number : 62423-73-8
  • Molecular Weight : 243.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150-154
Synonym 3-Carboxyphenacyl bromide, 2-Bromo-1-(3-carboxyphenyl)ethan-1-one
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromoacetylbenzoic-acid

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromoacetylbenzoic-acid
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart