APO455830266 Apollo Scientific Ltd

5-Chloropyridine-2-carboxylic acid

  • MDL number MFCD04114192
  • Cas Number : 86873-60-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 166-171
Synonym 2-Carboxy-5-chloropyridine, 5-Chloropicolinic acid
Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/5-chloropyridine-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-chloropyridine-2-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart